Methyl orange 0,05% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

R4980000-500A R4980000-120A
RC498016EA 82.43 CAD
RC498016 RC49804
Methyl orange 0,05% (w/v) in aqueous solution indicator
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: 90
Flash Pt: 38
MDL Number: MFCD00007502
CAS Number: 547-58-0
UN: 3143
ADR: 6.1 III
Merck Index: 13,06127

Order Now

Specification Test Results

Appearance Clear, orange, odorless
Suitability for Use Yellow (pH 4.4) - Pinkish-Orange (pH 3.2) - Pink (pH 3.2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR