Methylene blue (Loeffler) for staining and differentiation of acid fast organisms in smears
Supplier: Ricca Chemical
|
Danger
|
R4880000-500A
R4880000-120A
RC488016EA
115.13
CAD
RC488016
RC48804
Methylene blue (Loeffler) for staining and differentiation of acid fast organisms in smears
Methylene blue
Methylene blue, Loeffler formulation
Formula:
C₁₆H₁₈ClN₃S Boiling Pt: ~ 82 °C Melting Pt: ~ 0 °C Density: 0.9800 g/cm³ Flash Pt: 24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 UN: 1987 ADR: 3,III |
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Appearance | Dark blue, slight alcohol odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...