Methylene blue 1% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

R4850000-500A R4850000-120A R4850000-1A
RC485016EA 109.4 CAD
RC485016 RC48504 RC4850-32
Methylene blue 1% (w/v) in aqueous solution
Methylene blue
Formula: C₁₆H₁₈ClN₃S
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR