Methylene blue 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

R4841000-4C R4841000-120C
RC4841-1EA 1314.56 CAD
RC4841-1 RC4841-4
Methylene blue 0.1% (w/v) in aqueous solution
Methylene blue
Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
Merck Index: 12,06137

Order Now

Specification Test Results

Appearance Clear, blue, odorless
UV/Visible Spectrum Conforms to control


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR