Manganese(II) sulfate solution 364 g/L

Supplier: Ricca Chemical

Synonyms: Manganese sulphate, Manganous sulphate, Manganese sulfate, Manganous sulfate

R4620000-500A R4620000-1A
RC462016EA 140.62 CAD
RC462016 RC462032
Manganese(II) sulfate solution 364 g/L
Manganese (II) sulphate
Formula: MnO₄S
MW: 151
Density: 3.25
MDL Number: MFCD00003464
CAS Number: 7785-87-7
UN: 3077
ADR: 9 III
Merck Index: 12,05782

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 364 ± 4 g/L MnSO4.H2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR