Magnesium chloride 51% (w/v) in aqueous solution

Supplier: Ricca Chemical

R4470000-500A R4470000-1A
RC447016EA 230.68 CAD
RC447016 RC447032
Magnesium chloride 51% (w/v) in aqueous solution
Magnesium chloride
Formula: MgCl₂
Boiling Pt: ~ 100 °C
Density: 1.1791 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (by Refractive Index) 50.5-51.5 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR