Lactic acid 1.00 N

Supplier: Ricca Chemical
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

R4280000-500A R4280000-1A
RC428016EA 164.32 CAD
RC428016 RCR4280000-1A
Lactic acid 1.00 N
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless to slightly yellow liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 0.999-1.001 N at 20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR