Lactic acid 10% (v/v) in aqueous solution

Supplier: Ricca Chemical
Danger

R4270000-500A R4270000-4A
RC427016EA 104.28 CAD
RC427016 RC4270-1
Lactic acid 10% (v/v) in aqueous solution
Lactic acid
Formula: H₃CCH(OH)COOH
Boiling Pt: ~ 100…122 °C
Melting Pt: ~ 0 °C
Density: 1.0217 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless to slightly yellow liquid
Refractive Index Confirmed concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR