Kovac's indole reagent for detection of indole producing bacteria
Supplier: Ricca Chemical
|
Danger
|
R4260000-500C
R4260000-120C
RC426016EA
260.7
CAD
RC426016
RC42604
Kovac's indole reagent for detection of indole producing bacteria
Kovac's indole reagent
Kovac's Aldehyde Reagent, for Detection of Indole Producing Bacteria
Formula:
(CH₃)₂NC₆H₄CHO Boiling Pt: ~ 110 °C Density: 0.9525 g/cm³ Flash Pt: ~ 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003381 CAS Number: 100-10-7 UN: 2920 ADR: 8 (3),II |
Specification Test Results
Appearance | Clear, greenish-yellow, alcohol odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...