Ionic strength adjustor (ISA) for pH indicating and adjusting, for use with ammonia selective electrode
Supplier: Ricca Chemical
R4128000-500A
R4128000-1A
RC412816EA
123.24
CAD
RC412816
RC412832
Ionic strength adjustor (ISA) for pH indicating and adjusting, for use with ammonia selective electrode
Ionic strength adjustor (ISA)
Ionic strength adjustor, pH indicating and adjusting, for use with Ammonia Selective Electrode.
Specification Test Results
Appearance | Dark blue, methanol odor |
ppm Ammonia | No detectable Ammonia |