Hydroxylammonium chloride 50 g/ 100 mL in water
Supplier: Ricca Chemical
Synonyms:
Hydroxylamine HCl, Hydroxylamine hydrochloride
R3925000-500A
R3925000-120A
RC392516EA
508.76
CAD
RC392516
RC39254
Hydroxylammonium chloride 50 g/ 100 mL in water
Hydroxylammonium chloride
Formula:
NH₂OH·HCl MW: 69.49 g/mol Boiling Pt: 306 Melting Pt: 151 |
MDL Number:
MFCD00051089 CAS Number: 5470-11-1 UN: 2923 ADR: 8 III Merck Index: 13,04853 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...