Hydrogen sulfide saturated solution in water

Supplier: Ricca Chemical

R3850000-500C R3850000-1C
RC385016EA 557.74 CAD
RC385016 RC385032
Hydrogen sulfide saturated solution in water
Hydrogen sulphide
Formula: H₂S
CAS Number: 7783-06-4
UN: 1053
ADR: 2,

Order Now

Specification Test Results

Appearance Clear, colorless, strong sulfide odor
Suitability for Use Yellow precipitate - Saturation Test with Ferric Chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR