Hydrochloric acid 0.250 N (N/4) in isopropanol (IPA)

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3803000-20F R3803000-4A R3803000-1A
RC3803-5EA 1246.62 CAD
RC3803-5 RC3803-1 RC3803-32
Hydrochloric acid 0.250 N (N/4) in isopropanol (IPA)
Hydrochloric acid
Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, HCl/IPA odor
Assay at 20 °C (traceable to NIST SRM 84) 0.2500 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR