Hydrochloric acid 0.200 N (N/5) in isopropanol (IPA)

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3802000-4A R3802000-1A R3802000-20F
RC3802-1EA 462.94 CAD
RC3802-1 RC3802-32 RC3802-5
Hydrochloric acid 0.200 N (N/5) in isopropanol (IPA)
Hydrochloric acid
Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, IPA odor
Assay at 20 °C (traceable to NIST SRM 84) 0.2000 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR