Hydrochloric acid 2.50 N

Supplier: Ricca Chemical

R3715000-1A R3715000-4A
RC3715-32EA 88.48 CAD
RC3715-32 RC3715-1
Hydrochloric acid 2.50 N
Hydrochloric acid
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, slight Hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 2.500 ± 0.001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR