Hydrochloric acid 0.05882 N (N/17)

Supplier: Ricca Chemical

R3598000-1A R3598000-4A R3598000-500A
RC3598-32EA 71.1 CAD
RC3598-32 RC3598-1 RC3598-16
Hydrochloric acid 0.05882 N (N/17)
Hydrochloric acid
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.05882 ± 0.00005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR