Hydrochloric acid ≥75% (v/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3580750-4A R3580750-500A R3580750-1A
RC3580.75-1EA 434.5 CAD
RC3580.75-1 RC3580.75-16 RC3580.75-32
Hydrochloric acid ≥75% (v/v) in aqueous solution
Hydrochloric acid
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless to slight yellow-green, Hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 75.0 ± 0.5 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR