Hydrochloric acid 8,33% v/v aqueous solution

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3569400-500A R3569400-1A
RC3569.4-16EA 364.98 CAD
RC3569.4-16 RC3569.4-32
Hydrochloric acid 8,33% v/v aqueous solution
Hydrochloric acid
Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 8.33 ± 0.06 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR