Hydrochloric acid 1,5% v/v aqueous solution

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3565000-4A R3565000-500A R3565000-1A
RC3565-1EA 462.94 CAD
RC3565-1 RC3565-16 RC3565-32
Hydrochloric acid 1,5% v/v aqueous solution
Hydrochloric acid
Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 1.50 ± 0.04 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR