Hydrochloric acid, dilute R 73 g/L
Supplier: Ricca Chemical
|
Danger
|
R3556000-1A
R3556000-500A
RC3556-32EA
2518.52
CAD
RC3556-32
RC3556-16
Hydrochloric acid, dilute R 73 g/L
Hydrochloric acid, dilute R
Formula:
ClH Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.1500 g/cm³ Storage Temperature: Ambient |
UN:
1789 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, hydrochloric odor |
Assay at 20 °C (traceable to NIST SRM 84) | 73.0 ± 0.2 g/L HCl |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...