Hematoxylin solution Gill III

Supplier: Ricca Chemical
Danger

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

R3537000-4A R3537000-500A R3537000-1A
RC35371EA 807.38 CAD
RC35371 RC353716 RC353732
Hematoxylin solution Gill III
Hematoxylin staining solutions

Hematoxylin Stain Solution, Gill 3 Formulation, Triple Strength, for Frozen or Plastic Imbedded Sections, and for More Intense Nuclei in Progressive Histology Staining

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark purple, slight acetic acid odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR