Hematoxylin solution_SC Gill II

Supplier: Ricca Chemical
Danger

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

R3536000-4A R3536000-500A R3536000-1A R3536000-20F
RC35361EA 847.02 CAD
RC35361 RC353616 RC353632 RC3536-5
Hematoxylin solution_SC Gill II
Hematoxylin staining solutions

Hematoxylin Stain Solution, Gill 2 Formulation, Double Strength, for More Intense Nuclei in Progressive Cytology Staining and for Staining Delicate Nuclei in Progressive Histology Staining Procedures

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark purple, slight acetic acid odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR