Hematoxylin solution Gill I
Supplier: Ricca Chemical
|
Danger
|
R3535000-4A
R3535000-500A
R3535000-1A
RC35351EA
777.36
CAD
RC35351
RC353516
RC353532
Hematoxylin solution Gill I
Hematoxylin staining solutions
Hematoxylin Stain Solution, Gill 1 Formulation, Regular Strength, for Staining Delicate Nuclei in Progressive Cytology Staining Procedures
Formula:
C₁₆H₁₄O₆ Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0000 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Appearance | Dark purple, slight acetic acid odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...