Hematoxylin modified solution (Harris) for histology, for cytology mercury-free

Supplier: Ricca Chemical
Danger

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

R3530000-4C R3530000-500C R3530000-1C
RC35301EA 583.02 CAD
RC35301 RC353016 RC353032
Hematoxylin modified solution (Harris) for histology, for cytology mercury-free
Hematoxylin staining solutions

Nuclear Stain for General Histology and Cytology

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Boiling Pt: ~ 98 °C
Melting Pt: ~ 0 °C
Density: 1.0220 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark purple, odorless
Specific Gravity 1.022 ± 0.002 sp gr at 25.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR