Gram's iodine solution dilute Lugol's iodine for Gram staining
Supplier: Ricca Chemical
|
Warning
|
3450-1
3450-16
3450-32
RC34501EA
374.82
CAD
RC34501
RC345016
RC345032
Gram's iodine solution dilute Lugol's iodine for Gram staining
Gram's iodine solution
Formula:
I₃K Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0100 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00134128 CAS Number: 12298-68-9 |
Specification Test Results
Appearance | Dark brown with Iodine odor |
Assay at 20 °C (traceable to NIST SRM 136) | 0.026 ± 0.002 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...