Crystal violet 2% (w/v) in methanol

Supplier: Ricca Chemical

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

R3235000-500A R3235000-120A
RC323516EA 183.28 CAD
RC323516 RC32354
Crystal violet 2% (w/v) in methanol
Crystal Violet
Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) ABS ~590 nm
Appearance Violet purple with alcohol odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR