Crystal violet 1% (w/v) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
R3230000-500A
R3230000-120A
RC323016EA
129.64
CAD
RC323016
RC32304
Crystal violet 1% (w/v) in aqueous solution
Crystal Violet
Formula:
C₂₅H₃₀ClN₃ MW: 407.99 g/mol |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 UN: 3077 ADR: 9,III |
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Appearance | Clear, dark violet, odorless |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...