Iron(III) nitrate solution 404 g/L in dilute nitric acid
Supplier: Ricca Chemical
|
Danger
|
R3135000-500B
R3135000-1B
RC3135-16EA
456.62
CAD
RC3135-16
RC3135-32
Iron(III) nitrate solution 404 g/L in dilute nitric acid
Iron(III) nitrate solutions
Formula:
Fe(NO₃)₃·9H₂O Boiling Pt: ~ 100 °C Density: 1.2500 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 UN: 3264 ADR: 8,III |
Specification Test Results
Appearance | Clear, pale yellow, slightly pungent odor |
Assay at 20 °C (traceable to NIST SRM 136) | 404.0 ± 0.4 g/L |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...