Ferric chloride ≥100% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R3130000-500A
R3130000-120A
RC313016EA
518.24
CAD
RC313016
RC31304
Ferric chloride ≥100% (w/v) in aqueous solution
Iron(III) chloride solutions
Formula:
FeCl₃·6H₂O Boiling Pt: ~ 105 °C Density: 1.4700 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149712 CAS Number: 10025-77-1 UN: 2582 ADR: 8,III |
Specification Test Results
Appearance | Clear, brown, odorless |
Assay | 100 ± 3 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...