Ammonium iron (III) sulfate saturated solution in water

Supplier: Ricca Chemical
Warning

R3070000-500A R3070000-1A R3070000-120A
RC307016EA 202.24 CAD
RC307016 RC307032 RC30704
Ammonium iron (III) sulfate saturated solution in water
Ammonium iron(III) sulphate solutions
Formula: NH₄Fe(SO₄)₂·12H₂O
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7

Order Now

Specification Test Results

Appearance Dark brown, odorless
Suitability for Use White precipitate (Excess Silver ions) - Reddish-Brown Color (Excess Thiocyante ions)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR