Ferric alum, Volhard indicator
Supplier: Ricca Chemical
|
Danger
|
R3050000-500A
R3050000-120A
RC305016EA
221.2
CAD
RC305016
RC30504
Ferric alum, Volhard indicator
Ferric alum, Volhard
Formula:
FeH₂₈NO₂₀S₂ Boiling Pt: ~ 101 °C Density: 1.0800 g/cm³ Storage Temperature: Ambient |
UN:
3264 ADR: 8,III |
Specification Test Results
Appearance | Clear, gold, odorless |
Suitability for Use | White precipitate (Excess Silver ions) - Reddish-Brown Color (Excess Thiocyante ions) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...