Eosin Y (yellowish) 0.5% (w/v) in aqueous solution USP Test Solution (TS) adsorption indicator for argentometric titrations

Supplier: Ricca Chemical

R2858000-500A R2858000-120A
RC2858-16EA 150.1 CAD
RC2858-16 RC2858-4
Eosin Y (yellowish) 0.5% (w/v) in aqueous solution USP Test Solution (TS) adsorption indicator for argentometric titrations
Eosin Y (yellowish)
Formula: C₂₀H₆Br₄Na₂O₅
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1

Order Now

Specification Test Results

Appearance Clear, red, odorless
Suitability for Use Magenta (excess Silver ions)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR