Ehrlich aldehyde reagent solution alcoholic solution for indole test
Supplier: Ricca Chemical
|
Danger
|
R2790000-500C
R2790000-120C
RC279016EA
199.08
CAD
RC279016
RC27904
Ehrlich aldehyde reagent solution alcoholic solution for indole test
Ehrlich aldehyde reagent
Boiling Pt:
~ 80 °C Density: 0.9000 g/cm³ Storage Temperature: Ambient |
CAS Number:
100-10-7 UN: 2924 ADR: 3 (8),II |
Specification Test Results
Appearance | Clear, greenish-yellow, slightly pungent to pungent odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...