EDTA disodium salt 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Edetic acid disodium salt dihydrate, Titriplex III, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Edetate disodium dihydrate

R2670000-500A R2670000-120A
RC267016EA 140.62 CAD
RC267016 RC26704
EDTA disodium salt 10% (w/v) in aqueous solution
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6

Order Now

Specification Test Results

Appearance Colorless to slight yellow liquid
Assay at 20 °C (traceable to NIST SRM 915) 10.0 ± 0.4 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR