Diphenylcarbazone-xylene cyanol mixed indicator, for low level chloride determination, (not acidifier)
Supplier: Ricca Chemical
|
Danger
|
R2620000-500C
R2620000-120C
RC262016EA
421.86
CAD
RC262016
RC26204
Diphenylcarbazone-xylene cyanol mixed indicator, for low level chloride determination, (not acidifier)
Diphenylcarbazone-xylene cyanol
Boiling Pt:
~ 77 °C Density: 0.8000 g/cm³ Flash Pt: ~ 15.5 °C Storage Temperature: Refrigerator |
UN:
1987 ADR: 3,II |
Specification Test Results
Appearance | Purplish-blue to bluish green, alcohol odor |
Suitability for Use | Light Green (pH 3.8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...