Acid alcohol 5% HCl (v/v) in 95% denatured alcohol, strong
Supplier: Ricca Chemical
|
Danger
|
R0260000-4A
R0260000-500A
R0260000-1A
RC2601EA
360.24
CAD
RC2601
RC26016
RC26032
Acid alcohol 5% HCl (v/v) in 95% denatured alcohol, strong
Acid alcohol
Boiling Pt:
~ 82 °C Density: 0.8088 g/cm³ Flash Pt: ~ 23.5 °C Storage Temperature: Ambient |
UN:
2924 ADR: 3 (8),II |
Specification Test Results
Appearance | Clear, colorless, strong alcohol odor |
Assay at 20 °C (traceable to NIST SRM 84) | 5.0 ± 0.1 % (v/v) at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...