Acid alcohol 3% HCl in 70% alcohol Orth's decolorizing fluid
Supplier: Ricca Chemical
|
Danger
|
R0250000-4A
R0250000-500A
R0250000-1A
RC2501EA
462.94
CAD
RC2501
RC25016
RC25032
Acid alcohol 3% HCl in 70% alcohol Orth's decolorizing fluid
Acid alcohol
Boiling Pt:
~ 93 °C Melting Pt: ~ 0 °C Density: 0.8000 g/cm³ Flash Pt: ~ 23.5 °C Storage Temperature: Ambient |
UN:
1987 ADR: 3,II |
Specification Test Results
Appearance | Clear, colorless, alcohol odor |
Assay at 20 °C (traceable to NIST SRM 84) | 3.0 ± 0.1 % (v/v) at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...