Cuprous chloride reagent, acid carbon monoxide absorbent for Orsat gas analysis

Supplier: Ricca Chemical
Danger

R2500000-500C R2500000-1C
RC250016EA 153.26 CAD
RC250016 RC250032
Cuprous chloride reagent, acid carbon monoxide absorbent for Orsat gas analysis
Cuprous chloride reagent, acid
Formula: ClCu
MW: 99.9 g/mol
Boiling Pt: ~ 102 °C
Density: 1.1600 g/cm³
Storage Temperature: Ambient
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, dark greenish-brown, pungent odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR