Copper sulfate 1.055 Specific Gravity at 25°C (77°F) for hemoglobin estimation in blood donor screening of men

Supplier: Ricca Chemical
Danger

R2340000-4C R2340000-500C R2340000-1C R2340000-20F
RC23401EA 214.88 CAD
RC23401 RC234016 RC234032 RC2340-5
Copper sulfate 1.055 Specific Gravity at 25°C (77°F) for hemoglobin estimation in blood donor screening of men
Copper(II) sulphate solutions
Formula: CuSO₄·5H₂O
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0550 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8

Order Now

Specification Test Results

Appearance Clear, blue, odorless
Specific Gravity 1.0551 ± 0.0001 sp gr at 25.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR