Copper sulfate 2% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Copper(II) sulfate, Copper(II) sulphate, Cupric sulphate, Cupric sulfate
R2317000-1A
RC2317-32EA
124.82
CAD
RC2317-32
Copper sulfate 2% (w/v) in aqueous solution
Copper(II) sulphate solutions
Formula:
CuSO₄·5H₂O MW: 249.69 g/mol Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0150 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 |
Specification Test Results
Appearance | Clear, blue, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 2.0 ± 0.1 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...