Congo red 0,1% w/v aqueous solution Indicator

Supplier: Ricca Chemical
Danger

Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28

R2250000-120A R2250000-500A
RC22504EA 67.59 CAD
RC22504 RC2250-16
Congo red 0,1% w/v aqueous solution Indicator
Congo Red
Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004028
CAS Number: 573-58-0
Merck Index: 12,02562

Order Now

Specification Test Results

Appearance Clear, red, odorless
Suitability for Use Red (pH 5.0) - Blue (pH 3.0)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR