Conditioning reagent for sulfate analysis (turbidimetric)
Supplier: Ricca Chemical
|
Danger
|
R2235000-500A
R2235000-1A
RC223516EA
173.8
CAD
RC223516
RC223532
Conditioning reagent for sulfate analysis (turbidimetric)
Conditioning reagent
Boiling Pt:
~ 95 °C Density: 1.1272 g/cm³ Storage Temperature: Ambient |
UN:
3264 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, slight IPA odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...