Citric acid 1.00 M
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
R2131000-500A
R2131000-1A
R2131000-4A
RC2131-16EA
158
CAD
RC2131-16
RC2131-32
RCR2131000-4A
Citric acid 1.00 M
Citric acid
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 0.999-1.001 Molar |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...