Citrate-cyanide reducing solution for lead determination
Supplier: Ricca Chemical
|
Danger
|
R2112000-1A
R2112000-500A
RC2112-32EA
137.46
CAD
RC2112-32
RC2112-16
Citrate-cyanide reducing solution for lead determination
Citrate-cyanide reducing solution
Boiling Pt:
~ 100 °C Density: 1.0900 g/cm³ Storage Temperature: Ambient |
UN:
2672 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless to slight color, ammonia odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...