Cerium(IV) sulfate solution 0.100 N (N/10) in sulphuric acid 2N

Supplier: Ricca Chemical

Synonyms: Ceric sulfate, Cerium disulfate, Cerium disulphate, Ceric sulphate, Cerium(IV) sulfate, Cerium(IV) sulphate

R1900000-20F R1900000-4A R1900000-1A R1900000-500A
RC1900-5EA 1489.94 CAD
RC1900-5 RC19001 RC190032 RC1900-16
Cerium(IV) sulfate solution 0.100 N (N/10) in sulphuric acid 2N
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MDL Number: MFCD00148852
CAS Number: 13590-82-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, orange, odorless
Assay at 20 °C (traceable to NIST SRM 40) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR