Buffer Solution, Giordano, pH 6.4 (M/15), for Wright Staining, Ricca Chemical Company
Supplier: Ricca Chemical
R1450000-4A R1450000-500A R1450000-1A
RC14501EA 205.4 CAD
RC14501 RC145016 RC145032
Buffer Solution, Giordano, pH 6.4 (M/15), for Wright Staining, Ricca Chemical Company
Buffers Biological Buffers
For In-Vitro Diagnostic Use. FDA Registered Class Medical Device. Suitable for use in Wright, Wright-Giemsa, and Giemsa staining. This product may also be suitable for use in other methods.
Order Now
SPECIFICATIONS
Environmentally Preferable