Acetic acid 11.1% (v/v) in aqueous solution (1 + 8)

Supplier: Ricca Chemical

R0137000-500A R0137000-1A
RC137-16EA 363.4 CAD
RC137-16 RC137-32
Acetic acid 11.1% (v/v) in aqueous solution (1 + 8)
Acetic acid
Formula: H₃CCOOH
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, acetic odor
Assay at 20 °C (traceable to NIST SRM 84) 11.10 ± 0.05 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR