Bromocresol purple 0.1% (w/v) in dilute sodium hydroxide USP Test Solution (TS)
Supplier: Ricca Chemical
Synonyms:
5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 5',5''-Dibromo-o-cresolsulfonphthalein
R1310000-500A
R1310000-120A
RC1310-16EA
134.3
CAD
RC1310-16
RC1310-4
Bromocresol purple 0.1% (w/v) in dilute sodium hydroxide USP Test Solution (TS)
Bromocresol purple
Formula:
C₂₁H₁₆Br₂O₅S MW: 540.23 g/mol Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0010 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011681 CAS Number: 115-40-2 Merck Index: 13,01371 |
Specification Test Results
Appearance | Clear, purple, odorless |
Suitability for Use | Yellow (pH 6.8) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...