Bromocresol green-methyl red solution alcoholic solution mixed indicator

Supplier: Ricca Chemical
Danger

Synonyms: Methyl Red - Bromocresol green solution

R1220000-500A R1220000-120A
RC122016EA 221.66 CAD
RC122016 RC12204
Bromocresol green-methyl red solution alcoholic solution mixed indicator
Bromocresol green-methyl red
Boiling Pt: ~ 93 °C
Density: 0.9000 g/cm³
Flash Pt: ~ 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00134126
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Dark green, alcohol odor
Suitability for Use Greenish-Blue (pH 5.4) - Light Pink (pH 4.5), with Gray warning color

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR