Bromocresol green-methyl red in aqueous solution mixed indicator
Supplier: Ricca Chemical
Synonyms:
Methyl Red - Bromocresol green solution
R1215000-500A
R1215000-1A
R1215000-120A
RC1215-16EA
154.84
CAD
RC1215-16
RC1215-32
RC1215-4
Bromocresol green-methyl red in aqueous solution mixed indicator
Bromocresol green-methyl red
Specification Test Results
Appearance | Dark green-blue, odorless |
Suitability for Use | Greenish-Blue (pH 5.4) - Light Pink (pH 4.5), with Gray warning color |